5-Formyl-6-methoxypyridine-3-boronic acid pinacol ester ≥97%
Supplier: AOBChem USA
|
Warning
|
35040-1G
35040-25G
35040-500MG
35040-5G
76931-574EA
292.81
USD
76931-574
76931-576
76931-578
76931-580
5-Formyl-6-methoxypyridine-3-boronic acid pinacol ester ≥97%
5-Formyl-6-methoxypyridine-3-boronic acid pinacol ester
Precaución: for Research Use Only
Formula:
C₁₃H₁₈BNO₄ Storage Temperature: Refrigerator |
MDL Number:
MFCD08063052 CAS Number: 1310404-57-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...