(5-Bromo-2,3-difluoro-4-methoxyphenyl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 5-Bromo-2,3-difluoro-4-methoxyphenylboronic acid

34808-1G 34808-25G 34808-5G
76931-520EA 94.43 USD
76931-520 76931-522 76931-524
(5-Bromo-2,3-difluoro-4-methoxyphenyl)boronic acid ≥97%
(5-Bromo-2,3-difluoro-4-methoxyphenyl)boronic acid
Formula: C₇H₆BBrF₂O₃
MW: 266.83 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18914514
CAS Number: 2096337-64-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR