4-Fluoro-3,5-dimethylbenzyl alcohol ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (4-Fluoro-3,5-dimethylphenyl)methanol

34770-25G 34770-5G
76931-494EA 431.79 USD
76931-494 76931-496
4-Fluoro-3,5-dimethylbenzyl alcohol ≥97%
4-Fluoro-3,5-dimethylbenzyl alcohol

Precaución: for Research Use Only

Formula: C₉H₁₁FO
MW: 154.181 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06660267
CAS Number: 886501-76-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR