5,6-Dimethylnicotinic acid ≥95%

Supplier: AOBChem USA

Warning

34730-1G 34730-25G 34730-500MG 34730-5G
76931-482EA 155.96 USD
76931-482 76931-484 76931-486 76931-488
5,6-Dimethylnicotinic acid ≥95%
5,6-Dimethylnicotinic acid
Formula: C₈H₉NO₂
Storage Temperature: Refrigerator
MDL Number: MFCD13181797
CAS Number: 757903-81-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR