5-Chloro-2,4-difluorophenylboronic acid pinacol ester ≥97%
Supplier: AOBChem USA
|
Warning
|
34725-1G
34725-25G
34725-5G
76931-468EA
79.57
USD
76931-468
76931-470
76931-472
5-Chloro-2,4-difluorophenylboronic acid pinacol ester ≥97%
5-Chloro-2,4-difluorophenylboronic acid pinacol ester
Precaución: for Research Use Only
Formula:
C₁₂H₁₄BClF₂O₂ Storage Temperature: Refrigerator |
MDL Number:
MFCD08669564 CAS Number: 1073354-65-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...