5-Bromo-3-chloro-2-fluorophenol ≥97%

Supplier: AOBChem USA

Warning

34659-25G 34659-5G
76931-438EA 321.46 USD
76931-438 76931-440
5-Bromo-3-chloro-2-fluorophenol ≥97%
5-Bromo-3-chloro-2-fluorophenol
Formula: C₆H₃BrClFO
Storage Temperature: Refrigerator
MDL Number: MFCD18914516
CAS Number: 1305322-97-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR