Ethyl 4-fluoro-2-methylbenzoate ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-Fluoro-2-methylbenzoic acid ethyl ester

34577-1G 34577-25G 34577-5G
76931-426EA 73.21 USD
76931-426 76931-428 76931-430
Ethyl 4-fluoro-2-methylbenzoate ≥97%
Ethyl 4-fluoro-2-methylbenzoate
Formula: C₁₀H₁₁FO₂
MW: 182.19 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06204529
CAS Number: 167758-88-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR