4-Nitro-3-trifluoromethylphenylboronic acid, pinacol ester ≥97%

Supplier: AOBChem USA

Warning

34475-1G 34475-500MG 34475-5G
76931-376EA 498.63 USD
76931-376 76931-378 76931-380
4-Nitro-3-trifluoromethylphenylboronic acid, pinacol ester ≥97%
4-Nitro-3-trifluoromethylphenylboronic acid, pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₅BF₃NO₄
MW: 317.069 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD16996361
CAS Number: 1402615-44-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR