5,6-Dibromo-2-pyridinamine ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 5,6-Dibromopyridin-2-amine

34354-25G 34354-5G
76931-366EA 396.78 USD
76931-366 76931-368
5,6-Dibromo-2-pyridinamine ≥97%
5,6-Dibromo-2-pyridinamine

Precaución: for Research Use Only

Formula: C₅H₄Br₂N₂
MW: 251.907 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00234045
CAS Number: 89284-11-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR