3-Ethoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 5-Ethoxypyridine-3-boronic acid pinacol ester

34251-1G 34251-25G 34251-500MG 34251-5G
76931-354EA 185.66 USD
76931-354 76931-356 76931-358 76931-360
3-Ethoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine ≥97%
3-Ethoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Formula: C₁₃H₂₀BNO₃
MW: 249.11 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11878280
CAS Number: 1171892-40-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR