5-Bromo-2-fluoro-3-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

34139-1G 34139-25G 34139-500MG 34139-5G
76931-342EA 242.95 USD
76931-342 76931-344 76931-346 76931-348
5-Bromo-2-fluoro-3-methylbenzoic acid ≥97%
5-Bromo-2-fluoro-3-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₈H₆BrFO₂
MW: 233.034 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD23712061
CAS Number: 1427382-02-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR