5-p-Tolylpyridin-2-ylamine ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 5-(p-Tolyl)pyridin-2-amine, 5-(4-Methylphenyl)pyridin-2-amine

34046-1G 34046-25G 34046-5G
76931-328EA 62.6 USD
76931-328 76931-330 76931-332
5-p-Tolylpyridin-2-ylamine ≥97%
5-p-Tolylpyridin-2-ylamine

Precaución: for Research Use Only

Formula: C₁₂H₁₂N₂
MW: 184.240 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06797790
CAS Number: 503536-74-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR