4-Chloro-3-fluoro-2-methylphenol ≥97%

Supplier: AOBChem USA

Warning

33838-1G 33838-500MG 33838-5G
76931-282EA 558.04 USD
76931-282 76931-284 76931-286
4-Chloro-3-fluoro-2-methylphenol ≥97%
4-Chloro-3-fluoro-2-methylphenol

Precaución: for Research Use Only

Formula: C₇H₆ClFO
Storage Temperature: Refrigerator
MDL Number: MFCD28752138
CAS Number: 1804404-46-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR