5,8-Dibromoisoquinoline ≥97%

Supplier: AOBChem USA

Warning

33823-25G 33823-5G
76931-270EA 418 USD
76931-270 76931-272
5,8-Dibromoisoquinoline ≥97%
5,8-Dibromoisoquinoline

Precaución: for Research Use Only

Formula: C₉H₅Br₂N
MW: 286.95 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03109888
CAS Number: 81045-39-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR