4-Chloro-3-fluoro-2-hydroxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

33729-1G 33729-25G 33729-500MG 33729-5G
76931-232EA 290.69 USD
76931-232 76931-234 76931-236 76931-238
4-Chloro-3-fluoro-2-hydroxybenzaldehyde ≥97%
4-Chloro-3-fluoro-2-hydroxybenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₄ClFO₂
MW: 174.557 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22415101
CAS Number: 1427431-22-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR