2-Methoxy-p-toluic acid ≥97%

Supplier: AOBChem USA

Warning

33463-25G 33463-5G
76931-206EA 233.4 USD
76931-206 76931-208
2-Methoxy-p-toluic acid ≥97%
2-Methoxy-p-toluic acid

Precaución: for Research Use Only

Formula: C₉H₁₀O₃
Storage Temperature: Ambient
MDL Number: MFCD00020380
CAS Number: 704-45-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR