4-Fluoronaphtalene-1-boronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

33450-1G 33450-25G 33450-5G
76931-196EA 62.6 USD
76931-196 76931-198 76931-200
4-Fluoronaphtalene-1-boronic acid pinacol ester ≥97%
4-Fluoronaphtalene-1-boronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₆H₁₈BFO₂
MW: 272.122 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11520119
CAS Number: 627526-35-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR