4-Chloro-3-(trifluoromethoxy)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

33377-25G 33377-5G
76931-176EA 516.66 USD
76931-176 76931-178
4-Chloro-3-(trifluoromethoxy)benzaldehyde ≥97%
4-Chloro-3-(trifluoromethoxy)benzaldehyde

Precaución: for Research Use Only

Formula: C₈H₄ClF₃O₂
MW: 224.57 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06660219
CAS Number: 886499-59-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR