4-Methoxy-2,3-dimethylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2,3-Dimethylanisaldehyde

33339-25G 33339-5G
76931-168EA 233.4 USD
76931-168 76931-170
4-Methoxy-2,3-dimethylbenzaldehyde ≥97%
4-Methoxy-2,3-dimethylbenzaldehyde
Formula: C₁₀H₁₂O₂
MW: 164.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003339
CAS Number: 38998-17-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR