5-Bromo-2-chloro-4-fluorobenzoic acid ≥97%

Supplier: AOBChem USA

Warning

33275-1G 33275-25G 33275-500MG 33275-5G
76931-136EA 272.66 USD
76931-136 76931-138 76931-140 76931-142
5-Bromo-2-chloro-4-fluorobenzoic acid ≥97%
5-Bromo-2-chloro-4-fluorobenzoic acid
Formula: C₇H₃BrClFO₂
MW: 253.453 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD24107203
CAS Number: 1204219-98-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR