4-Fluoro-3-methoxy-5-methylbenzoic acid ≥95%

Supplier: AOBChem USA

Warning

33236-1G 33236-500MG 33236-5G
76931-124EA 524.09 USD
76931-124 76931-126 76931-128
4-Fluoro-3-methoxy-5-methylbenzoic acid ≥95%
4-Fluoro-3-methoxy-5-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₉H₉FO₃
MW: 184.164 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD15527528
CAS Number: 1427418-74-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR