(4'-Methoxy-biphenyl-4-yl)methanol ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 4'-Methoxy-[1,1'-biphenyl]-4-methanol

33005-1G 33005-25G 33005-500MG 33005-5G
76931-104EA 272.66 USD
76931-104 76931-106 76931-108 76931-110
(4'-Methoxy-biphenyl-4-yl)methanol ≥95%
(4'-Methoxy-biphenyl-4-yl)methanol

Precaución: for Research Use Only

Formula: C₁₄H₁₄O₂
MW: 214.26 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD01862509
CAS Number: 20854-60-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR