4-Bromo-2-ethoxybenzyl alcohol ≥97%

Supplier: AOBChem USA

Warning

32967-1G 32967-25G 32967-5G
76931-096EA 96.55 USD
76931-096 76931-098 76931-100
4-Bromo-2-ethoxybenzyl alcohol ≥97%
4-Bromo-2-ethoxybenzyl alcohol

Precaución: for Research Use Only

Formula: C₉H₁₁BrO₂
Storage Temperature: Ambient
MDL Number: MFCD11652000
CAS Number: 1094750-85-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR