5-Fluoro-4-methoxy-3-(trifluoromethyl)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

32936-1G 32936-25G 32936-500MG 32936-5G
76931-082EA 318.27 USD
76931-082 76931-084 76931-086 76931-088
5-Fluoro-4-methoxy-3-(trifluoromethyl)benzoic acid ≥97%
5-Fluoro-4-methoxy-3-(trifluoromethyl)benzoic acid

Precaución: for Research Use Only

Formula: C₉H₆F₄O₃
MW: 238.136 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD31805758
CAS Number: 2244107-74-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR