4'-Methoxybiphenyl-4-carboxaldehyde ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4'-Methoxy-[1,1'-biphenyl]-4-carbaldehyde

32928-25G 32928-5G
76931-072EA 421.18 USD
76931-072 76931-074
4'-Methoxybiphenyl-4-carboxaldehyde ≥97%
4'-Methoxybiphenyl-4-carboxaldehyde
Formula: C₁₄H₁₂O₂
MW: 212.248 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD01862517
CAS Number: 52988-34-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR