4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-2-(trifluoromethyl)phenol ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-Hydroxy-3-trifluoromethylphenylboronic acid pinacol ester

32915-1G 32915-25G 32915-500MG 32915-5G
76931-058EA 173.99 USD
76931-058 76931-060 76931-062 76931-064
4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-2-(trifluoromethyl)phenol ≥97%
4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-2-(trifluoromethyl)phenol
Formula: C₁₃H₁₆BF₃O₃
MW: 288.07 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16994250
CAS Number: 1243143-45-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR