4-Chloro-2,5-difluorophenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-(4-Chloro-2,5-difluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

32807-1G 32807-25G 32807-500MG 32807-5G
76931-044EA 221.73 USD
76931-044 76931-046 76931-048 76931-050
4-Chloro-2,5-difluorophenylboronic acid pinacol ester ≥97%
4-Chloro-2,5-difluorophenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₂H₁₄BClF₂O₂
MW: 274.499 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22418270
CAS Number: 1126320-24-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR