5-Bromo-2-methoxy-3-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

32731-1G 32731-25G 32731-500MG 32731-5G
76931-012EA 231.28 USD
76931-012 76931-014 76931-016 76931-018
5-Bromo-2-methoxy-3-methylbenzaldehyde ≥97%
5-Bromo-2-methoxy-3-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₉H₉BrO₂
Storage Temperature: Refrigerator
MDL Number: MFCD26059145
CAS Number: 145742-37-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR