5-Bromo-2-methoxy-4-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

32723-1G 32723-25G 32723-500MG 32723-5G
76931-000EA 290.69 USD
76931-000 76931-002 76931-004 76931-006
5-Bromo-2-methoxy-4-methylbenzaldehyde ≥97%
5-Bromo-2-methoxy-4-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₉H₉BrO₂
Storage Temperature: Refrigerator
MDL Number: MFCD21234728
CAS Number: 923281-67-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR