4-Methoxy-2-(propan-2-yloxy)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

32560-1G 32560-25G 32560-500MG 32560-5G
76930-962EA 162.32 USD
76930-962 76930-964 76930-966 76930-968
4-Methoxy-2-(propan-2-yloxy)benzaldehyde ≥97%
4-Methoxy-2-(propan-2-yloxy)benzaldehyde

Precaución: for Research Use Only

Formula: C₁₁H₁₄O₃
MW: 194.227 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD14607625
CAS Number: 107811-48-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR