4-Fluoro-3-methoxy-5-methylphenylboronic acid pinacol ester ≥97%
Supplier: AOBChem USA
|
Warning
|
32404-1G
32404-25G
32404-500MG
32404-5G
76930-938EA
290.69
USD
76930-938
76930-940
76930-942
76930-944
4-Fluoro-3-methoxy-5-methylphenylboronic acid pinacol ester ≥97%
4-Fluoro-3-methoxy-5-methylphenylboronic acid pinacol ester
Precaución: for Research Use Only
Formula:
C₁₄H₂₀BFO₃ Storage Temperature: Ambient |
MDL Number:
MFCD30529810 CAS Number: 2121514-30-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...