5-Bromo-2,4-dimethoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

32331-25G 32331-5G
76930-882EA 315.09 USD
76930-882 76930-884
5-Bromo-2,4-dimethoxybenzoic acid ≥97%
5-Bromo-2,4-dimethoxybenzoic acid
Formula: C₉H₉BrO₄
MW: 261.071 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05865172
CAS Number: 32246-20-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR