4-Chloro-2-fluoro-3-methoxyphenol ≥97%

Supplier: AOBChem USA

Warning

32214-1G 32214-25G 32214-500MG 32214-5G
76930-856EA 409.51 USD
76930-856 76930-858 76930-860 76930-862
4-Chloro-2-fluoro-3-methoxyphenol ≥97%
4-Chloro-2-fluoro-3-methoxyphenol

Precaución: for Research Use Only

Formula: C₇H₆ClFO₂
Storage Temperature: Refrigerator
MDL Number: MFCD30344685
CAS Number: 1993479-29-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR