4-Bromo-1,3-thiazole-2-carboxylic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-Bromothiazole-2-carboxylic acid

32189-25G 32189-5G
76930-848EA 381.93 USD
76930-848 76930-850
4-Bromo-1,3-thiazole-2-carboxylic acid ≥97%
4-Bromo-1,3-thiazole-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₄H₂BrNO₂S
MW: 208.033 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09701413
CAS Number: 88982-82-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR