2-(4-Chloro-2,3-difluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-Chloro-2,3-difluorophenylboronic acid pinacol ester

32170-1G 32170-25G 32170-5G
76930-806EA 117.76 USD
76930-806 76930-808 76930-810
2-(4-Chloro-2,3-difluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97%
2-(4-Chloro-2,3-difluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Formula: C₁₂H₁₄BClF₂O₂
MW: 274.50 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD16659094
CAS Number: 1116681-98-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR