4'-Fluoro-3'-methoxyacetophenone ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 1-(4-Fluoro-3-methoxyphenyl)ethanone

32056-25G 32056-5G
76930-782EA 439.22 USD
76930-782 76930-784
4'-Fluoro-3'-methoxyacetophenone ≥97%
4'-Fluoro-3'-methoxyacetophenone

Precaución: for Research Use Only

Formula: C₉H₉FO₂
Storage Temperature: Ambient
MDL Number: MFCD00272135
CAS Number: 64287-19-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR