5-Chloropyridine-2-carboxamide ≥97%

Supplier: AOBChem USA

Warning

31960-25G 31960-5G
76930-762EA 240.83 USD
76930-762 76930-764
5-Chloropyridine-2-carboxamide ≥97%
5-Chloropyridine-2-carboxamide

Precaución: for Research Use Only

Formula: C₆H₅ClN₂O
MW: 156.57 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10574693
CAS Number: 370104-72-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR