2-(4-Ethoxy-3-ethylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-Ethoxy-3-ethylphenylboronic acid pinacol ester

31761-1G 31761-500MG 31761-5G
76930-738EA 736.27 USD
76930-738 76930-740 76930-742
2-(4-Ethoxy-3-ethylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97%
2-(4-Ethoxy-3-ethylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Formula: C₁₆H₂₅BO₃
MW: 276.1789 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD30344676
CAS Number: 2121514-94-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR