5-Bromo-2-ethoxyanisole ≥97%

Supplier: AOBChem USA

Warning

31645-1G 31645-25G 31645-5G
76930-712EA 91.24 USD
76930-712 76930-714 76930-716
5-Bromo-2-ethoxyanisole ≥97%
5-Bromo-2-ethoxyanisole

Precaución: for Research Use Only

Formula: C₉H₁₁BrO₂
Storage Temperature: Ambient
MDL Number: MFCD18837618
CAS Number: 1070795-38-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR