5-Acetylthiophene-3-boronic acid pinacol ester ≥95%

Supplier: AOBChem USA

Warning

31440-1G 31440-25G 31440-500MG 31440-5G
76930-690EA 439.22 USD
76930-690 76930-692 76930-694 76930-696
5-Acetylthiophene-3-boronic acid pinacol ester ≥95%
5-Acetylthiophene-3-boronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₂H₁₇BO₃S
Storage Temperature: Refrigerator
MDL Number: MFCD12198153
CAS Number: 1219628-58-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR