5-Bromo-2-methoxynicotinonitrile ≥95%

Supplier: AOBChem USA

Warning

31268-25G 31268-5G
76930-656EA 453.01 USD
76930-656 76930-658
5-Bromo-2-methoxynicotinonitrile ≥95%
5-Bromo-2-methoxynicotinonitrile

Precaución: for Research Use Only

Formula: C₇H₅BrN₂O
Storage Temperature: Refrigerator
MDL Number: MFCD09801047
CAS Number: 941294-54-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR