4-Iodo-2-methoxynicotinic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-Iodo-2-methoxypyridine-3-carboxylic acid

31223-1G 31223-25G 31223-500MG 31223-5G
76930-640EA 272.66 USD
76930-640 76930-642 76930-644 76930-646
4-Iodo-2-methoxynicotinic acid ≥97%
4-Iodo-2-methoxynicotinic acid

Precaución: for Research Use Only

Formula: C₇H₆INO₃
Storage Temperature: Ambient
MDL Number: MFCD26520771
CAS Number: 726206-55-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR