4-Chloro-3-hydroxyphenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Chloro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol

31150-1G 31150-25G 31150-500MG 31150-5G
76930-582EA 270.53 USD
76930-582 76930-584 76930-586 76930-588
4-Chloro-3-hydroxyphenylboronic acid pinacol ester ≥97%
4-Chloro-3-hydroxyphenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₂H₁₆BClO₃
MW: 254.518 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD16994405
CAS Number: 1443151-85-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR