(4-Chloro-2-(methoxycarbonyl)phenyl)boronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 4-Chloro-2-(methoxycarbonyl)phenylboronic acid

31101-25G 31101-5G
76930-554EA 601.54 USD
76930-554 76930-556
(4-Chloro-2-(methoxycarbonyl)phenyl)boronic acid ≥97% research grade
(4-Chloro-2-(methoxycarbonyl)phenyl)boronic acid
Formula: C₈H₈BClO₄
MW: 214.4100 g/mol
Storage Temperature: Ambient
MDL Number: MFCD25459257
CAS Number: 1612256-37-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR