5-Fluoro-2-(methoxycarbonyl)benzeneboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: Methyl 4-fluoro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate, 5-Fluoro-2-(methoxycarbonyl)phenylboronic acid pinacol ester

31081-1G 31081-25G 31081-500MG 31081-5G
76930-540EA 120.95 USD
76930-540 76930-542 76930-544 76930-546
5-Fluoro-2-(methoxycarbonyl)benzeneboronic acid pinacol ester ≥97%
5-Fluoro-2-(methoxycarbonyl)benzeneboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₄H₁₈BFO₄
MW: 280.10 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD16996304
CAS Number: 1400976-17-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR