4-Chloro-3-methoxyphenol ≥97%

Supplier: AOBChem USA

Warning

31021-1G 31021-25G 31021-5G
76930-512EA 106.09 USD
76930-512 76930-514 76930-516
4-Chloro-3-methoxyphenol ≥97%
4-Chloro-3-methoxyphenol

Precaución: for Research Use Only

Formula: C₇H₇ClO₂
MW: 158.582 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD16998742
CAS Number: 18113-07-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR