4-Methoxy-3-(1-methylethyl)phenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

31018-1G 31018-25G 31018-500MG 31018-5G
76930-498EA 290.69 USD
76930-498 76930-500 76930-502 76930-504
4-Methoxy-3-(1-methylethyl)phenylboronic acid pinacol ester ≥97%
4-Methoxy-3-(1-methylethyl)phenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₆H₂₅BO₃
Storage Temperature: Ambient
MDL Number: MFCD22414615
CAS Number: 2121513-08-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR