2-(4-Chloro-3-fluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-Chloro-3-fluorophenylboronic acid pinacol ester, 4-Chloro-3-fluorobenzeneboronic acid pinacol ester

30992-1G 30992-25G 30992-5G
76930-488EA 73.21 USD
76930-488 76930-490 76930-492
2-(4-Chloro-3-fluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97%
2-(4-Chloro-3-fluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Formula: C₁₂H₁₅BClFO₂
MW: 256.508 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16996258
CAS Number: 627525-83-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR