5-Bromo-1,3-difluoro-2-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

30922-25G 30922-5G
76930-476EA 201.58 USD
76930-476 76930-478
5-Bromo-1,3-difluoro-2-methylbenzene ≥97%
5-Bromo-1,3-difluoro-2-methylbenzene
Formula: C₇H₅BrF₂
MW: 207.02 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07368697
CAS Number: 179617-08-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR