5-Bromo-6-methyl-2H-1,3-benzodioxole ≥97%

Supplier: AOBChem USA

Warning

30859-1G 30859-25G 30859-500MG 30859-5G
76930-448EA 393.6 USD
76930-448 76930-450 76930-452 76930-454
5-Bromo-6-methyl-2H-1,3-benzodioxole ≥97%
5-Bromo-6-methyl-2H-1,3-benzodioxole

Precaución: for Research Use Only

Formula: C₈H₇BrO₂
MW: 215.044 g/mol
Storage Temperature: Refrigerator
CAS Number: 5025-53-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR